Learn More
Sino Biological Recombinant Human CD52 Protein (hFc Tag)
Supplier:  Sino Biological 15105H02H100
Encompass_Preferred
A DNA sequence encoding the human CD52 (NP_001794.2) (Met1-Ser36) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9399