Learn More
Sino Biological Recombinant Human Defensin alpha 3/DEFA3 Protein (hFc Tag)
Supplier:  Sino Biological 15109H01H100
Encompass_Preferred
A DNA sequence encoding the human DEFA3 (NP_005208.1) (Asp39-Cys94) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-9401