Learn More
Sino Biological Recombinant Human TPPP2 Protein (His Tag)
Supplier:  Sino Biological 15112H07E100
Encompass_Preferred
A DNA sequence encoding the human TPPP2 (AAH38970.1) (Met1-Lys170) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9403