Learn More
Sino Biological Recombinant Human LRRC3B Protein (hFc Tag)
Supplier:  Sino Biological 15120H02H100
Encompass_Preferred
A DNA sequence encoding the human LRRC3B (NP_443185.1) (Met1-Tyr204) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9409