Learn More
Sino Biological Recombinant Human PRPS2 Protein (His Tag)
Supplier:  Sino Biological 15128H08E50
Encompass_Preferred
A DNA sequence encoding the human PRPS2 (P11908-1) (Met1-Leu318) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-9415