Learn More
Waters Corp WATERS CORPORATION Performance Maintenance Kit, 1515 Isocratic Pump, Annual
Catalog No. 50791545 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the 1515 isocratic pump contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. This kit is designed to ensure optimal functionality and longevity of your HPLC system.
  • Contains two Clear-100 seals to ensure proper functioning.
  • Includes two sapphire plungers for reliable operation.
  • Features four PerformancePLUS cartridges for consistent performance.
  • Comes with one reference valve rebuild kit for comprehensive maintenance.
  • One solvent filter is included for clean system operation.
  • Designed for annual replacement to maintain peak performance.
  • Suitable for HPLC system type applications.
  • Optimized for the 1515 isocratic pump.
  • Provided as one unit per package for convenience.
Catalog No. 50-791-545 Supplier Waters Corp Supplier No. 201000113
The Waters™ Performance Maintenance Kit for the 1515 isocratic pump contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. This kit is designed to ensure optimal functionality and longevity of your HPLC system.
  • Contains two Clear-100 seals to ensure proper functioning.
  • Includes two sapphire plungers for reliable operation.
  • Features four PerformancePLUS cartridges for consistent performance.
  • Comes with one reference valve rebuild kit for comprehensive maintenance.
  • One solvent filter is included for clean system operation.
  • Designed for annual replacement to maintain peak performance.
  • Suitable for HPLC system type applications.
  • Optimized for the 1515 isocratic pump.
  • Provided as one unit per package for convenience.