Learn More
Sino Biological Recombinant Human HNT/NTM Protein (hFc Tag)
Supplier:  Sino Biological 15169H02H100
Encompass_Preferred
A DNA sequence encoding the human NTM (NP_057606.1) (Met1-Gly312) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9424