Learn More
Sino Biological Recombinant Human Arp3/ACTR3 Protein (His & GST Tag)
Supplier:  Sino Biological 15179H20B100
Encompass_Preferred
A DNA sequence encoding the human ACTR3 (NP_005712.1) (Ala2- Ser418) was expressed with a N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9429