Learn More
Sino Biological Recombinant Human Pellino-1 Protein (His & GST Tag)
Supplier:  Sino Biological 15180H20B100
Encompass_Preferred
A DNA sequence encoding the human PELI1 (NP_065702.2) (Met1-Asp418) was expressed with a N-terminal GST-tagged polyhistidine tag at the N-terminus.
Catalog No. 50-161-9431