Learn More
Sino Biological Recombinant Human DBI Protein (His Tag)
Supplier:  Sino Biological 15211H07E100
Encompass_Preferred
A DNA sequence encoding the human DBI (P07108-2) (Met1-Ala87) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9439