Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, 1525 Binary Pump, Annual
Catalog No. 50791546 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the 1525 binary pump includes all the Waters Quality Parts™ necessary for annual maintenance to ensure the instrument operates at peak performance.
  • Contains all quality parts for annual maintenance
  • Designed for the 1525 binary pump
  • Includes four Clear-100 seals
  • Includes four sapphire plungers
  • Includes eight PerformancePLUS cartridges
  • Includes one reference valve rebuild kit
  • Includes two solvent filters
Catalog No. 50-791-546 Supplier Waters Corp Supplier No. 201000114
The Waters™ Performance Maintenance Kit for the 1525 binary pump includes all the Waters Quality Parts™ necessary for annual maintenance to ensure the instrument operates at peak performance.
  • Contains all quality parts for annual maintenance
  • Designed for the 1525 binary pump
  • Includes four Clear-100 seals
  • Includes four sapphire plungers
  • Includes eight PerformancePLUS cartridges
  • Includes one reference valve rebuild kit
  • Includes two solvent filters