Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, HPLC, Annual
Catalog No. 50791570 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the 1525EF pump contains all the necessary parts to keep your instrument operating at peak performance on an annual basis. The kit includes four plunger seals, four extended flow plungers, two extended flow check valve kits, two outlet check valve kits, one reference valve rebuild kit, and two solvent filters.
  • Designed for annual maintenance.
  • Helps maintain peak instrument performance.
  • Includes a comprehensive set of replacement parts: plunger seals, extended flow plungers, extended flow check valve kits, outlet check valve kits, reference valve rebuild kit, and solvent filters.
Catalog No. 50-791-570 Supplier Waters Corp Supplier No. 201000160
The Waters™ Performance Maintenance Kit for the 1525EF pump contains all the necessary parts to keep your instrument operating at peak performance on an annual basis. The kit includes four plunger seals, four extended flow plungers, two extended flow check valve kits, two outlet check valve kits, one reference valve rebuild kit, and two solvent filters.
  • Designed for annual maintenance.
  • Helps maintain peak instrument performance.
  • Includes a comprehensive set of replacement parts: plunger seals, extended flow plungers, extended flow check valve kits, outlet check valve kits, reference valve rebuild kit, and solvent filters.