Learn More
Waters Corp Plunger Seal, 1525 EF, 1 pk
Supplier:  Waters Corp 700002282
Encompass_Preferred
The 1525EF Plunger Seal is designed to maintain pressure within the pump head. It is crucial for preventing leaks, and replacement is necessary if the seal becomes damaged.
  • Helps maintain pressure in the pump head
  • Prevents leaking from the pump head
  • Replacement is required if damaged
  • Includes 1 unit per package
  • Categorized as a spare parts kit
Catalog No. 50-818-738