Learn More
Waters Corp Tubing Kit, Stainless Steel, HPLC, 1525EF Binary Pump
Catalog No. 502392753 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement set of stainless steel tubing assemblies designed for the 1525 Extended Flow Binary Pump. The kit includes all external tubing assemblies that may require replacement for optimal system performance.
  • Spare parts kit
  • For HPLC systems
  • Replacement as required
Catalog No. 50-239-2753 Supplier Waters Corp Supplier No. 700002679
This is a replacement set of stainless steel tubing assemblies designed for the 1525 Extended Flow Binary Pump. The kit includes all external tubing assemblies that may require replacement for optimal system performance.
  • Spare parts kit
  • For HPLC systems
  • Replacement as required