Learn More
Waters Corp Waters Performance Maintenance Kit, 1525 Micro, HPLC, Annual Replacement
Catalog No. 50791571 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the 1525 micro pump contains all the necessary parts to keep your instrument operating at peak performance on an annual basis.
  • Contains two seal kits
  • Includes four sapphire plungers
  • Features five check valves (package of 2)
  • Comes with one rotor seal
  • Equipped with two solvent filters
Catalog No. 50-791-571 Supplier Waters Corp Supplier No. 201000161
The Performance Maintenance Kit for the 1525 micro pump contains all the necessary parts to keep your instrument operating at peak performance on an annual basis.
  • Contains two seal kits
  • Includes four sapphire plungers
  • Features five check valves (package of 2)
  • Comes with one rotor seal
  • Equipped with two solvent filters