Learn More
Sino Biological Recombinant Human ALR Protein (His Tag)
Supplier:  Sino Biological 15278H07E100
Encompass_Preferred
A DNA sequence encoding the human GFER (P55789-2) (Met1-Asp125) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9455