Learn More
Sino Biological Recombinant Human RBP5 Protein (His Tag)
Supplier:  Sino Biological 15282H07E100
Encompass_Preferred
A DNA sequence encoding the human RBP5 (P82980) (Met1-Arg135) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9459