Learn More
Sino Biological Recombinant Human Dermcidin Protein (hFc Tag)
Supplier:  Sino Biological 15332H01H100
Encompass_Preferred
A DNA sequence encoding the human DCD (NP_444513.1) (Ser63-Val109) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-9473