Learn More
Sino Biological Recombinant Human LY6D Protein (mFc Tag)
Supplier:  Sino Biological 15334H05H100
Encompass_Preferred
A DNA sequence encoding the human LY6D (CAA73189.1) (Met1-His97) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9477