Learn More
Sino Biological Recombinant Human LACRT Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 15335H08H100
Encompass_Preferred
A DNA sequence encoding the human LACRT (NP_150593.1) (Met1-Ala138) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9479