Learn More
Sino Biological Recombinant Human CST8 Protein (hFc Tag)
Supplier:  Sino Biological 15336H02H100
Encompass_Preferred
A DNA sequence encoding the human CST8 (NP_005483.1) (Met1-Ala142) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9481