Learn More
Sino Biological Recombinant Human Olfactory Marker Protein Protein (His Tag)
Supplier:  Sino Biological 15345H07E100
Encompass_Preferred
A DNA sequence encoding the human OMP (P47874) (Met1-Leu163) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9485