Learn More
Aurora Microplates 1536 LOW BASE SQ WELL BLK PLAT
Catalog No. NC1509548
Encompass
Poly-D-Lysine coated Aurora 1536 Low Base, Square Well, black plate, 100 uM Film Bottom without lids, with barcodes15/bag, 60 per case
Catalog No. NC1509548 Supplier Aurora Microplates Supplier No. EBA141000BPDL
Add to Cart
Add to Cart
Poly-D-Lysine coated Aurora 1536 Low Base, Square Well, black plate, 100 uM Film Bottom without lids, with barcodes15/bag, 60 per case