Learn More
Corning™ 1536-Well Black/Clear Round Bottom Ultra-low Attachment Spheroid Microplate with Lid, Sterile
Shop All Corning Products

Description
- 1536-well format allows for high throughput 3D cell culture
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
- Unique design shields each well to minimize well-to-well cross-talk.
- You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
- Compatible with the GNF ultra high throughput system
Specifications
Specifications
| Barcode | No Barcode |
| Lid | With Lid |
| Array | 32 x 48 |
| Color | Black, Clear Bottom |
| Product Type | Spheroid Microplate |
| Volume (Metric) Well | 14 mL |
| No. of Wells | 1536 |
| Volume (Metric) Working | 5 to 10 μL |
| Sterility | Sterile |
| Surface Treatment | Ultra-low Binding |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.