Learn More
Greiner Bio-One 1536-well HiBase Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07000002 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
White
Surface Treatment:
FLUOTRAC 200
LUMITRAC 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Surface Treatment
07-000-002 Black FLUOTRAC 200
07-000-004 White LUMITRAC 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-002 Supplier Greiner Bio-One Supplier No. 782076
Add to Cart
Add to Cart
Suited for luminescence measurements. Greiner Bio-One 1536-Well HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Sample carryover and cross-contamination risk is minimized
  • Total
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description Plate design: HiBase
Array 32 x 48
Color Black
Material Polystyrene
No. of Wells 1536
Sterility Non-sterile
Surface Treatment FLUOTRAC 200
Well Shape Flat Bottom (F-well)
Quantity 60/Cs.