Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 1536-Well Nontreated Cyclic Olefin Copolymer Plates
GREENER_CHOICE

Catalog No. 12456704 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Quantity:
10/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
12-456-704 Black, Clear Bottom 10/Pk.
12-456-700 Black, Clear Bottom 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-456-704 Supplier Corning™ Supplier No. 4565
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonsterile microplates have smooth top and clear, flat bottom.

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Format Standard
Lid Without Lid
Culture Area 0.03 cm2
Array 32 x 48
Binding Type Not Treated
Color Black, Clear Bottom
Product Type 1536-Well Assay Microplate
For Use With (Equipment) Automated dispensers
Material Cycloolefin Copolymer
Volume (Metric) Well 12.5 μL
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
For Use With (Application) For higher sensitivity
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 10/Pk.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.