Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 1536-Well Nontreated Polystyrene Plates
GREENER_CHOICE

Catalog No. 07202510 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White
Volume (Metric) Working:
8 μL
10 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Working
07-202-510 Black 10 μL
07-202-511 White 8 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-202-510 Supplier Corning™ Supplier No. 3724
Add to Cart
Add to Cart

Superior flatness minimizes variability within a plate and between plates

  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer™ FlexDrop™
  • Working volume of up to 10 μL
Generic Bar Code
  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate No sequence number duplicates

Order Info

Packaged in bags of 10

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
Color Black
For Use With (Equipment) Automated dispensers
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 10 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For barcode scanning and software systems
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.