Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
GREENER_CHOICE
Catalog No. 07200873 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
White, Clear Bottom
Includes:
Barcode
Generic barcodes
Without barcode
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
07-200-873 Black, Clear Bottom Without barcode
07-201-09 Black Generic barcodes
12-456-710 White, Clear Bottom Barcode
12-456-706 Black, Clear Bottom Barcode
05-400-01 Black Without barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-200-873 Supplier Corning™ Supplier No. 3895
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 32 x 48
Color Black, Clear Bottom
Product Type 1536-Well Cell Culture Multiwell Plates
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-binding
Total Volume (Metric) Well 12.5 μL
Well Shape Flat Bottom (F-well)
Includes Without barcode
No. per Case 50/Cs.
No. per Pack 10/Pk.
Well Color Black