Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 1536-Well SCREENSTAR Microplate with Lid

Catalog No. 07001197 Shop All Greiner Bio-One Products
Change view
Click to view available options
Total Volume (Metric) Well:
17.8 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Total Volume (Metric) Well
07-001-197 17.8 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-001-197 Supplier Greiner Bio-One Supplier No. 789866691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Superior Optical Clarity Supports High-Quality Imaging and Cellular Analysis

  • Ultra-Low Base Design Optimized for High Magnification Microscopy Applications
  • Excellent Plate Flatness Helps Reduce Autofocusing Time During Image Acquisition
  • Smooth Plate Surface Supports Automated Handling and Sealing Workflows
  • Black Frame with Clear Bottom Helps Improve Image Contrast
  • Tissue Culture Treated Surface Supports Cell Culture Applications
  • 1536-Well Format Designed for High-Content and High-Throughput Screening
TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Format SBS Standard
Lid Yes
Culture Area 2.1 mm2
Array 32 x 48
Color Black, Clear Bottom
Product Type Microplate
Material Advanced Polymer
No. of Wells 1536
Volume (Metric) Working 3 to 15 μL
Sterility Sterile
Surface Treatment Tissue Culture
Total Volume (Metric) Well 17.8 μL
Well Shape Square, Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) High Throughput and High Content Screening
High-throughput Compatibility Yes
No. per Case 48/Cs.
No. per Pack 12/Pk.
RNase Free Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.