Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 1536-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
GREENER_CHOICE

Catalog No. 07200799 Shop All Corning Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07-200-799 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-799 Supplier Corning™ Supplier No. 4527
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.

  • 1536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
  • Compatible with the GNF ultra high throughput system
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid With Lid
Array 32 x 48
Packaging 10/Pk., 50/Cs.
Binding Type Ultra-Low Attachment
Color Black, Clear Bottom
Product Type Spheroid
Material Polystyrene
Volume (Metric) Well 14 μL
No. of Wells 1536
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round Bottom (U-well)
For Use With (Application) 3D Cell Culture
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color Clear
Length (Metric) 125.75 mm
Width (Metric) 84.12 mm
Height (Metric) 8 mm
Depth (Metric) 6.1 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.