Learn More
Corning™ 1536-Well White Polystyrene NBS Microplate without Lid, Non-sterile
Catalog No. 07001142 Shop All Corning Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-001-142 White
1 options
Catalog No. 07-001-142 Supplier Corning™ Supplier No. 3729
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
  • NBS microplates are ideal for homogeneous assays with enzymes or for reducing nonspecific binding to microplate wells
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Color White
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working Up to 8 μL
Sterility Non-sterile
Surface Treatment NBS
Quantity 10/Pk., 50/Cs.