Learn More
Corning™ 1536-Well White Polystyrene Not Treated Microplate, No Lid, With Generic Bar Code, Non-sterile
GREENER_CHOICE
Catalog No. 07201456 Shop All Corning Products
Change view
Click to view available options
No. of Wells:
1536
Catalog No. No. of Wells
07-201-456 1536
1 options
Catalog No. 07-201-456 Supplier Corning™ Supplier No. 3725BC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Generic bar coded Corning microplates are now available in stock and ready to ship. Code 128 bar code format ensures compatibility with most bar code scanning and software systems. To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface).
  • Labels applied to all 4 sides of the plate
  • No sequence number duplicates
  • Exceptional print quality and resolution
  • Resistant to commonly used laboratory reagents
  • Resistant to temperature ranges of -80ºC to +52ºC
Microplate Features:
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness -minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200 ng IgG/cm²)
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Lid Without Lid
No. of Wells 1536
Volume (Metric) Working Up to 8 μL
Sterility Non-sterile
Surface Treatment Untreated
Quantity 10/Pk., 50/Cs.