Learn More
Sino Biological Recombinant Human DCUN1D2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 15363H07E100
Encompass_Preferred
A DNA sequence encoding the human DCUN1D2 (NP_001014305.1) (Met1-Phe259) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9489