Learn More
Aurora Microplates 1536K WELL BLK/CLR ST/TR 60/CS
Catalog No. 50245327
Encompass
Black, Aurora 1536 LoBase SQ, 100u, TC Treated-CO2, Sterile, No Lid, Kalypsys Bar Code, Standard Pack, 60 Plates Per Case
Catalog No. 50-245-327 Supplier Aurora Microplates Supplier No. ABI141001A
Black, Aurora 1536 LoBase SQ, 100u, TC Treated-CO2, Sterile, No Lid, Kalypsys Bar Code, Standard Pack, 60 Plates Per Case