Learn More
Sino Biological Recombinant Human APCDD1 Protein (hFc Tag)
Supplier:  Sino Biological 15383H02H100
Encompass_Preferred
A DNA sequence encoding the human APCDD1 (NP_694545.1) (Met1-Gly486) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9501