Learn More
Sino Biological Recombinant Human GPIHBP1 Protein (hFc Tag)
Supplier:  Sino Biological 15388H02H100
Encompass_Preferred
A DNA sequence encoding the human GPIHBP1 (AAH63857.1) (Met1-Ser160) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9503