Learn More
Sino Biological Recombinant Human GRIK2 Protein (hFc Tag)
Supplier:  Sino Biological 15398H02H100
Encompass_Preferred
A DNA sequence encoding the human GRIK2 (NP_001159719.1) (Met1-Pro561) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9514