Learn More
Sino Biological Recombinant Human STC1 Protein (His Tag)
Supplier:  Sino Biological 15411H08H100
Encompass_Preferred
A DNA sequence encoding the human STC1 (NP_003146.1) (Met1-Ala247) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9516