Learn More
Sino Biological Recombinant Human GGPS1 Protein (His Tag)
Supplier:  Sino Biological 15417H08H100
Encompass_Preferred
A DNA sequence encoding the human GGPS1 (NP_001032354.1) (Met1-Glu300) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9518