Learn More
Sino Biological Recombinant Human PARM-1 Protein (His Tag)
Supplier:  Sino Biological 15420H08H100
Encompass_Preferred
A DNA sequence encoding the human PARM1 (AAH13294.1) (Met1-Ser258) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9520