Learn More
Sino Biological Recombinant Human Annexin A8 Protein (His Tag)
Supplier:  Sino Biological 15421H07E100
Encompass_Preferred
A DNA sequence encoding the human ANXA8 (AAH73755.1) (Met1-Pro327) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9522