Learn More
Sino Biological Recombinant Human PFDN5 Protein (His Tag)
Supplier:  Sino Biological 15442H07E100
Encompass_Preferred
A DNA sequence encoding the human PFDN5 (NP_002615.2) (Met1-Ala154) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9542