Learn More
Sino Biological Recombinant Human Neurotensin Protein (hFc Tag)
Supplier:  Sino Biological 15443H02H100
Encompass_Preferred
A DNA sequence encoding the human NTS (NP_006174.1) (Met1-Leu148) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9544