Learn More
Sino Biological Recombinant Human MRAP Protein (hFc Tag)
Supplier:  Sino Biological 15444H01H100
Encompass_Preferred
A DNA sequence encoding the human MRAP (NP_848932.1) (Tyr59-Ser172) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-9546