Learn More
Sino Biological Recombinant Human RAB1B Protein (hFc Tag)
Supplier:  Sino Biological 15447H01H100
Encompass_Preferred
A DNA sequence encoding the human RAB1B (NP_112243.1) (Met1-Gly199) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-9548