Learn More
Sino Biological Recombinant Human GSTM5 Protein (His Tag)
Supplier:  Sino Biological 15453H07E100
Encompass_Preferred
A DNA sequence encoding the human GSTM5 (AAH58881.1) (Met1-Lys218) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9550