Learn More
Sino Biological Recombinant Human FNDC4 Protein (ECD, mFc Tag)
Supplier:  Sino Biological 15459H05H100
Encompass_Preferred
A DNA sequence encoding the human FNDC4 (NP_073734.1) (Met1-Thr167) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9552