Learn More
Sino Biological Recombinant Human FAM20B Protein (hFc Tag)
Supplier:  Sino Biological 15465H01H100
Encompass_Preferred
A DNA sequence encoding the human FAM20B (NP_055679.1)(Ser31-Leu409) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-9556