Learn More
Sino Biological Recombinant Human Endothelin B Receptor Protein (mFc Tag)
Supplier:  Sino Biological 15473H05H100
Encompass_Preferred
A DNA sequence encoding the human EDNRB (NP_000106.1) (Met1-Lys101) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9558