Learn More
Sino Biological Recombinant Human Tapasin Protein (His Tag)
Supplier:  Sino Biological 15478H08H100
Encompass_Preferred
A DNA sequence encoding the human TAPBP (AAH80574.1) (Met1-Val414) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9562